Assay Details
Target Gene Details
Entrez Gene ID: | 14158 |
Gene Name: | fer (fms/fps related) protein kinase |
Gene Aliases: |
AV082135, C330004K01Rik, Fert, Fert2 |
Location: |
Chr.17:63829950-64146519 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fer | NM_001037997.3 | 3 | 425 | NP_001033086.2 |
| NM_001286415.1 | NP_001273344.1 | |||
| AF286537.1 | 2 | 291 | AAG40730.1 | |
| AK021175.1 | 3 | 402 | BAB32315.1 | |
| AK157828.1 | 3 | 392 | BAE34213.1 | |
| AK165973.1 | 3 | 426 | BAE38493.1 | |
| BC058100.1 | AAH58100.1 | |||
| U76762.1 | 2 | 358 | AAB18988.1 |

Genomic Map