Assay Details
Target Gene Details
Entrez Gene ID: | 76416 |
Gene Name: | Znrd1 antisense |
Gene Aliases: |
1700022C21Rik, AI746295, Znrd1-as |
Location: |
Chr.17:36958592-36965625 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Znrd1as | NM_029602.1 | NP_083878.1 | ||
| AK006236.1 | BAB24473.1 | |||
| BC027007.1 | AAH27007.1 |

Genomic Map