Assay Details
Target Gene Details
Entrez Gene ID: | 240058 |
Gene Name: | copine V |
Gene Aliases: |
A830083G22Rik |
Location: |
Chr.17:29156521-29238118 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpne5 | NM_153166.2 | NP_694806.1 | ||
| AK044042.1 | BAC31750.1 | |||
| BC036971.1 | AAH36971.1 |
More Information
Set Membership: |
|

Genomic Map