Assay Details
Target Gene Details
Entrez Gene ID: | 11307 |
Gene Name: | ATP-binding cassette, sub-family G (WHITE), member 1 |
Gene Aliases: |
AW413978, Abc8, White |
Location: |
Chr.17:31057694-31117984 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcg1 | NM_009593.2 | 2 | 290 | NP_033723.1 |
| AF323659.1 | 2 | 286 | AAK27442.1 | |
| AK037433.1 | 2 | 210 | ||
| BC119471.2 | 2 | 238 | ||
| BY296839.1 | 2 | 291 | ||
| U34920.1 | 2 | 248 | AAB47738.1 | |
| Z48745.1 | 2 | 277 | CAA88636.1 |
More Information
Set Membership: |
|

Genomic Map