Assay Details
Target Gene Details
Entrez Gene ID: | 69192 |
Gene Name: | DEAH (Asp-Glu-Ala-His) box polypeptide 16 |
Gene Aliases: |
2410006N22Rik, DBP2, Ddx16, mKIAA0577 |
Location: |
Chr.17:35879764-35892670 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhx16 | NM_026987.2 | NP_081263.2 | ||
| AK122314.2 | BAC65596.4 | |||
| BC009147.1 | AAH09147.1 | |||
| BC046781.1 | ||||
| BC138204.1 | ||||
| BC138205.1 |
More Information
Set Membership: |
|

Genomic Map