Assay Details
Target Gene Details
Entrez Gene ID: | 75296 |
Gene Name: | Fgfr1 oncogene partner |
Gene Aliases: |
4930553O10Rik, FOP |
Location: |
Chr.17:8165485-8196477 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgfr1op | NM_001197046.1 | NP_001183975.1 | ||
| NM_201230.5 | NP_957682.1 | |||
| AK016110.1 | ||||
| AK028778.1 | BAC26115.1 | |||
| AK029470.1 | BAC26465.1 | |||
| AK031261.1 | BAC27326.1 | |||
| AK050389.1 | BAC34230.1 | |||
| BB615739.1 | ||||
| BC080717.1 | AAH80717.1 | |||
| BC108331.1 | AAI08332.1 |

Genomic Map