Assay Details
Target Gene Details
Entrez Gene ID: | 75296 |
Gene Name: | Fgfr1 oncogene partner |
Gene Aliases: |
4930553O10Rik, FOP |
Location: |
Chr.17:8165485-8196477 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgfr1op | NM_001197046.1 | 5 | 451 | NP_001183975.1 |
| NM_201230.5 | 5 | 451 | NP_957682.1 | |
| AK016110.1 | 5 | 454 | ||
| AK028778.1 | 5 | 380 | BAC26115.1 | |
| AK029470.1 | 5 | 450 | BAC26465.1 | |
| AK031261.1 | 5 | 434 | BAC27326.1 | |
| AK050389.1 | 5 | 389 | BAC34230.1 | |
| BB615739.1 | 5 | 453 | ||
| BC080717.1 | 5 | 438 | AAH80717.1 | |
| BC108331.1 | 5 | 449 | AAI08332.1 |

Genomic Map