Assay Details
Target Gene Details
Entrez Gene ID: | 14229 |
Gene Name: | FK506 binding protein 5 |
Gene Aliases: |
D17Ertd592e, Dit1, FKBP-5, FKBP51 |
Location: |
Chr.17:28398753-28486149 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkbp5 | NM_010220.4 | 2 | 257 | NP_034350.1 |
| AK169795.1 | 2 | 198 | BAE41371.1 | |
| BC015260.1 | 2 | 196 | AAH15260.1 | |
| BY740772.1 | 2 | 257 | ||
| CA317455.1 | 2 | 220 | ||
| CT010372.1 | 1 | 38 | CAJ18579.1 | |
| U16959.1 | 2 | 257 | AAA86983.1 | |
| U36220.1 | 2 | 185 | AAA89162.1 |
More Information
Set Membership: |
|

Genomic Map