Assay Details
Target Gene Details
Entrez Gene ID: | 77128 |
Gene Name: | CREB3 regulatory factor |
Gene Aliases: |
A930001N09Rik, LRF |
Location: |
Chr.17:26714711-26780985 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crebrf | NM_029870.2 | NP_084146.1 | ||
| AK030092.1 | BAC26779.1 | |||
| AK132423.1 | ||||
| AK132454.1 | 7 | 1838 | ||
| BC113191.1 | ||||
| BC113192.1 |

Genomic Map