Assay Details
Target Gene Details
Entrez Gene ID: | 114584 |
Gene Name: | chloride intracellular channel 1 |
Gene Aliases: |
Clcp, G6 |
Location: |
Chr.17:35050243-35058719 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clic1 | NM_033444.2 | NP_254279.1 | ||
| AK088809.1 | BAC40585.1 | |||
| AK150382.1 | BAE29513.1 | |||
| AK150611.1 | BAE29702.1 | |||
| AK153509.1 | BAE32054.1 | |||
| AK167765.1 | BAE39798.1 | |||
| AK168262.1 | BAE40210.1 | |||
| AK186702.1 | ||||
| BC004658.1 | AAH04658.1 |
More Information
Set Membership: |
|

Genomic Map