Assay Details
Target Gene Details
Entrez Gene ID: | 81701 |
Gene Name: | EGF-like domain 8 |
Gene Aliases: |
Ng3 |
Location: |
Chr.17:34613200-34616572 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Egfl8 | NM_152922.3 | NP_690886.3 | ||
| AK148723.1 | BAE28647.1 | |||
| AY635583.1 | AAT47548.1 | |||
| BC139484.1 | ||||
| BC141312.1 | ||||
| BC145632.1 |
More Information
Set Membership: |
|

Genomic Map