Assay Details
Target Gene Details
Entrez Gene ID: | 14236 |
Gene Name: | forkhead box N2 |
Gene Aliases: |
3230402J05Rik, 6030465J18Rik, Fkh19, Foxn1, HTLF |
Location: |
Chr.17:88440712-88490533 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Foxn2 | NM_180974.4 | NP_851305.2 | ||
| AK030735.1 | BAC27104.1 | |||
| AK031639.1 | ||||
| AK154401.1 | ||||
| BC138824.1 | ||||
| BC138825.1 | ||||
| CB524750.1 |

Genomic Map