Assay Details
Target Gene Details
Entrez Gene ID: | 224697 |
Gene Name: | a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 10 |
Gene Aliases: |
9430006O18, AW045948, Adam-ts10, Adamts-10, Znmp |
Location: |
Chr.17:33524176-33553782 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adamts10 | NM_172619.3 | NP_766207.2 | ||
| NR_037707.1 | ||||
| NR_037708.1 | ||||
| AK042525.1 | ||||
| AK048600.1 | ||||
| AK076334.1 | ||||
| BQ571453.1 |
More Information
Set Membership: |
|

Genomic Map