Assay Details
Target Gene Details
Entrez Gene ID: | 110147 |
Gene Name: | euchromatic histone lysine N-methyltransferase 2 |
Gene Aliases: |
Bat8, D17Ertd710e, G9a, KMT1C, NG36 |
Location: |
Chr.17:34898464-34914051 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ehmt2 | NM_001286573.1 | 22 | 3107 | NP_001273502.1 |
| NM_001286575.1 | 24 | 3045 | NP_001273504.1 | |
| NM_145830.2 | 23 | 3209 | NP_665829.1 | |
| NM_147151.2 | 23 | 2943 | NP_671493.1 | |
| AB077209.1 | 23 | 2936 | BAC05482.1 | |
| AB077210.1 | 23 | 3209 | BAC05483.1 | |
| AK077743.1 | 11 | 1265 | BAC36989.1 | |
| AK154246.1 | 22 | 3130 | BAE32462.1 | |
| AK191368.1 | 2 | 127 | ||
| AK204430.1 | 2 | 127 | ||
| BC025539.1 | 4 | 344 | AAH25539.1 | |
| BC058357.1 | 24 | 3043 | AAH58357.1 |

Genomic Map