Assay Details
Target Gene Details
Entrez Gene ID: | 26426 |
Gene Name: | nucleotide binding protein 2 |
Gene Aliases: |
D17Wsu11e |
Location: |
Chr.17:24882611-24886367 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nubp2 |
Target Gene Details
Entrez Gene ID: | 79043 |
Gene Name: | splA/ryanodine receptor domain and SOCS box containing 3 |
Gene Aliases: |
2310012N15Rik, 3300001M01Rik, SSB3, Tce1, mKIAA4204 |
Location: |
Chr.17:24886349-24892147 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spsb3 |

Genomic Map