Assay Details
Target Gene Details
Entrez Gene ID: | 26426 |
Gene Name: | nucleotide binding protein 2 |
Gene Aliases: |
D17Wsu11e |
Location: |
Chr.17:24882611-24886367 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nubp2 | NM_011956.3 | 3 | 292 | NP_036086.1 |
| AF114169.1 | 3 | 270 | AAF01785.1 | |
| AK003816.1 | 3 | 290 | BAB23013.1 | |
| AK169896.1 | 3 | 251 | ||
| AK210395.1 | 1 | 75 | ||
| AK213247.1 | 1 | 75 | ||
| BC012635.1 | 3 | 293 | AAH12635.1 | |
| BI735707.1 | 3 | 292 |

Genomic Map