Assay Details
Target Gene Details
Entrez Gene ID: | 106707 |
Gene Name: | RNA pseudouridylate synthase domain containing 1 |
Gene Aliases: |
2310051D06Rik, AU019540, Rlucl |
Location: |
Chr.17:25727703-25731464 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpusd1 | NM_028009.3 | 2 | 191 | NP_082285.1 |
| AK009926.1 | ||||
| AK049334.1 | 2 | 192 | BAC33690.1 | |
| BC018186.1 | 2 | 152 | AAH18186.1 |
More Information
Set Membership: |
|

Genomic Map