Assay Details
Target Gene Details
Entrez Gene ID: | 321007 |
Gene Name: | serine active site containing 1 |
Gene Aliases: |
4930511N22Rik, D17Ertd141e |
Location: |
Chr.17:6040571-6079762 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serac1 | NM_001111017.1 | NP_001104487.1 | ||
| NM_177311.4 | NP_796285.2 | |||
| AK019677.1 | BAB31833.1 | |||
| AK154605.1 | BAE32707.1 | |||
| AK155571.1 | BAE33329.1 | |||
| AK162246.1 | BAE36813.1 | |||
| AK170498.1 | BAE41837.1 | |||
| BC119594.1 | ||||
| BC119595.1 | ||||
| BY197974.1 | ||||
| BY715309.1 |

Genomic Map