Assay Details
Target Gene Details
Entrez Gene ID: | 224938 |
Gene Name: | praja 2, RING-H2 motif containing |
Gene Aliases: |
AI447901, AL022700, Neurodap1, mKIAA0438 |
Location: |
Chr.17:64281006-64331883 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pja2 | NM_001025309.1 | NP_001020480.1 | ||
| NM_144859.2 | NP_659108.1 | |||
| AF493070.1 | AAO85470.1 | |||
| AK122282.1 | BAC65564.1 | |||
| AK144586.1 | BAE25949.1 | |||
| AK168371.1 | BAE40303.1 | |||
| BC004742.1 | AAH04742.1 | |||
| BC017130.1 | AAH17130.1 | |||
| BC119446.1 |
More Information
Set Membership: |
|

Genomic Map