Assay Details
Target Gene Details
Entrez Gene ID: | 16004 |
Gene Name: | insulin-like growth factor 2 receptor |
Gene Aliases: |
AI661837, CD222, CI-MPR, M6P/IGF2R, Mpr300 |
Location: |
Chr.17:12682406-12769706 on Build GRCm38 |
Assay Gene Location: | Within Exon 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igf2r | NM_010515.2 | 35 | 5251 | NP_034645.2 |
| AK088699.1 | 10 | 1477 | BAC40512.1 | |
| BC031190.1 | 2 | 257 | AAH31190.1 | |
| BC150817.1 | 35 | 5162 | ||
| BC171973.1 | 35 | 5162 | ||
| BQ900123.1 | 4 | 578 | ||
| U04710.1 | 35 | 5207 | AAA19568.1 | |
| X60389.1 | 2 | 206 | CAA42940.1 |
More Information
Set Membership: |
|

Genomic Map