Assay Details
Target Gene Details
Entrez Gene ID: | 77596 |
Gene Name: | adhesion G protein-coupled receptor F1 |
Gene Aliases: |
5031409J19Rik, Gpr110, KPG_009 |
Location: |
Chr.17:43270276-43325876 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adgrf1 | NM_133776.2 | NP_598537.2 | ||
| AK052799.1 | BAC35151.1 | |||
| BB661727.1 | ||||
| BC019217.1 | AAH19217.1 |

Genomic Map