Assay Details
Target Gene Details
Entrez Gene ID: | 70530 |
Gene Name: | leucine rich repeat and fibronectin type III domain containing 2 |
Gene Aliases: |
5730420O05Rik, SALM1, mKIAA1246 |
Location: |
Chr.17:48931566-49097590 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrfn2 | NM_027452.3 | 1 | 38 | NP_081728.2 |
| AK017594.1 | 1 | 241 | BAB30828.1 | |
| AK122476.1 | BAC65758.1 | |||
| BC116651.1 | 1 | 200 | ||
| BC139418.1 | 1 | 38 |
More Information
Set Membership: |
|

Genomic Map