Assay Details
Target Gene Details
Entrez Gene ID: | 64657 |
Gene Name: | mitochondrial ribosomal protein S10 |
Gene Aliases: |
1110038B19Rik, AW743084, Rpms10 |
Location: |
Chr.17:47368887-47381418 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps10 | NM_001146211.1 | NP_001139683.1 | ||
| NM_001146212.1 | NP_001139684.1 | |||
| AK004151.1 | ||||
| AK142675.1 | ||||
| BB862036.1 | ||||
| DV046794.1 |
More Information
Set Membership: |
|

Genomic Map