Assay Details
Target Gene Details
Entrez Gene ID: | 435528 |
Gene Name: | glycine-N-acyltransferase-like 3 |
Gene Aliases: |
EG435528, Gm5683 |
Location: |
Chr.17:40904623-40920715 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glyatl3 | NM_001145060.1 | NP_001138532.1 |
More Information
Set Membership: |
|

Genomic Map