Assay Details
Target Gene Details
Entrez Gene ID: | 14651 |
Gene Name: | hydroxyacyl glutathione hydrolase |
Gene Aliases: |
BC019817, Glo-2, Glo2, Rsp29 |
Location: |
Chr.17:24840079-24864450 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hagh | NM_024284.2 | NP_077246.2 | ||
| AK147022.1 | BAE27615.1 | |||
| BC004749.1 | AAH04749.1 | |||
| BY348635.1 |

Genomic Map