Assay Details
Target Gene Details
Entrez Gene ID: | 14651 |
Gene Name: | hydroxyacyl glutathione hydrolase |
Gene Aliases: |
BC019817, Glo-2, Glo2, Rsp29 |
Location: |
Chr.17:24840079-24864450 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hagh | NM_001159626.1 | 5 | 498 | NP_001153098.1 |
| NM_024284.2 | 5 | 588 | NP_077246.2 | |
| AK147022.1 | 5 | 549 | BAE27615.1 | |
| AK150557.1 | 5 | 506 | BAE29657.1 | |
| AK151231.1 | 5 | 506 | BAE30223.1 | |
| AK152742.1 | 5 | 506 | BAE31462.1 | |
| AK165777.1 | 5 | 492 | BAE38377.1 | |
| BC004749.1 | 5 | 531 | AAH04749.1 | |
| BC019817.1 | 5 | 481 | AAH19817.1 | |
| BQ128470.1 |

Genomic Map