Assay Details
Target Gene Details
Entrez Gene ID: | 26416 |
Gene Name: | mitogen-activated protein kinase 14 |
Gene Aliases: |
CSBP2, Crk1, Csbp1, Mxi2, PRKM14, PRKM15, p38, p38-alpha, p38MAPK, p38a, p38alpha |
Location: |
Chr.17:28691342-28748405 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapk14 | NM_001168508.1 | 11 | 1290 | NP_001161980.1 |
| NM_001168513.1 | 11 | 891 | NP_001161985.1 | |
| NM_001168514.1 | 12 | 1335 | NP_001161986.1 | |
| NM_011951.3 | 11 | 1290 | NP_036081.1 | |
| AF195850.1 | 8 | 613 | AAF06348.1 | |
| AK089059.1 | 11 | 1204 | BAC40726.1 | |
| AK133684.1 | 11 | 1203 | BAE21782.1 | |
| AK151348.1 | 11 | 892 | BAE30324.1 | |
| AK153025.1 | 11 | 892 | BAE31659.1 | |
| BC012235.1 | 11 | 1290 | AAH12235.1 | |
| BC085303.1 | 11 | 1266 | AAH85303.1 | |
| CX568915.1 | 10 | 684 | ||
| D83073.1 | 11 | 1121 | BAA19741.1 | |
| U10871.1 | 11 | 1188 | AAA20888.1 |
More Information
Set Membership: |
|

Genomic Map