Assay Details
Target Gene Details
Entrez Gene ID: | 268930 |
Gene Name: | protein kinase, membrane associated tyrosine/threonine 1 |
Gene Aliases: |
6230424P17, AW209059, Myt1 |
Location: |
Chr.17:23726336-23736735 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pkmyt1 | NM_023058.3 | 5 | 1484 | NP_075545.2 |
| AF175892.1 | 5 | 1368 | AAG09210.1 | |
| AK031787.1 | 4 | 2351 | ||
| AK158550.1 | 5 | 1485 | BAE34554.1 | |
| AK165164.1 | 5 | 1351 | BAE38056.1 | |
| BC025061.1 | 5 | 1207 | AAH25061.1 |
More Information
Set Membership: |
|

Genomic Map