Assay Details
Target Gene Details
Entrez Gene ID: | 268930 |
Gene Name: | protein kinase, membrane associated tyrosine/threonine 1 |
Gene Aliases: |
6230424P17, AW209059, Myt1 |
Location: |
Chr.17:23726336-23736735 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pkmyt1 | NM_023058.3 | NP_075545.2 | ||
| AF175892.1 | AAG09210.1 | |||
| AK031787.1 | ||||
| AK158550.1 | BAE34554.1 | |||
| AK165164.1 | BAE38056.1 | |||
| BC025061.1 | AAH25061.1 |
More Information
Set Membership: |
|

Genomic Map