Assay Details
Target Gene Details
Entrez Gene ID: | 104215 |
Gene Name: | ras homolog gene family, member Q |
Gene Aliases: |
Arhq, TC10A, Tc10 |
Location: |
Chr.17:86963111-87000069 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhoq | NM_145491.2 | NP_663466.2 | ||
| AB060650.1 | BAB91068.1 | |||
| BC048813.1 | AAH48813.2 | |||
| BC056363.1 | AAH56363.1 |
More Information
Set Membership: |
|

Genomic Map