Assay Details
Target Gene Details
Entrez Gene ID: | 72900 |
Gene Name: | NADH dehydrogenase (ubiquinone) flavoprotein 2 |
Gene Aliases: |
2900010C23Rik |
Location: |
Chr.17:66078795-66101559 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufv2 | NM_001278415.1 | 6 | 579 | NP_001265344.1 |
| NM_028388.3 | 5 | 482 | NP_082664.1 | |
| AK013511.1 | 5 | 433 | BAB28888.1 | |
| AK078351.1 | 5 | 426 | BAC37233.1 | |
| AK088193.1 | 2 | 221 | BAC40201.1 | |
| AK146998.1 | 5 | 416 | BAE27595.1 | |
| AK150404.1 | 6 | 516 | BAE29530.1 | |
| AK151729.1 | 6 | 516 | BAE30647.1 | |
| AK159460.1 | 5 | 399 | BAE35102.1 | |
| AK166539.1 | 5 | 443 | BAE38841.1 | |
| AK169143.1 | 5 | 453 | BAE40922.1 | |
| AK185171.1 | 1 | 105 | ||
| BC030946.1 | 5 | 411 | AAH30946.1 | |
| BY246389.1 | ||||
| CF106105.1 | 4 | 324 |
More Information
Set Membership: |
|

Genomic Map