Assay Details
Target Gene Details
Entrez Gene ID: | 72900 |
Gene Name: | NADH dehydrogenase (ubiquinone) flavoprotein 2 |
Gene Aliases: |
2900010C23Rik |
Location: |
Chr.17:66078795-66101559 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufv2 | NM_001278415.1 | NP_001265344.1 | ||
| NM_028388.3 | NP_082664.1 | |||
| AK013511.1 | BAB28888.1 | |||
| AK078351.1 | BAC37233.1 | |||
| AK088193.1 | BAC40201.1 | |||
| AK146998.1 | BAE27595.1 | |||
| AK150404.1 | BAE29530.1 | |||
| AK151729.1 | BAE30647.1 | |||
| AK159460.1 | BAE35102.1 | |||
| AK166539.1 | BAE38841.1 | |||
| AK169143.1 | BAE40922.1 | |||
| AK185171.1 | ||||
| BC030946.1 | AAH30946.1 |
More Information
Set Membership: |
|

Genomic Map