Assay Details
Target Gene Details
Entrez Gene ID: | 78330 |
Gene Name: | NADH dehydrogenase (ubiquinone) flavoprotein 3 |
Gene Aliases: |
1500032D16Rik |
Location: |
Chr.17:31520115-31531326 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufv3 | NM_001083891.1 | NP_001077360.1 | ||
| NM_030087.2 | NP_084363.2 | |||
| AK077433.1 | BAC36798.1 | |||
| AK154191.1 | BAE32428.1 | |||
| AK154477.1 | BAE32613.1 | |||
| AK160289.1 | BAE35728.1 | |||
| BC013640.1 | AAH13640.1 | |||
| BU555486.1 | ||||
| BY064612.1 |
More Information
Set Membership: |
|

Genomic Map