Assay Details
Target Gene Details
Entrez Gene ID: | 23984 |
Gene Name: | phosphodiesterase 10A |
Gene Aliases: |
- |
Location: |
Chr.17:8525187-8986648 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde10a | NM_001290707.1 | 14 | 1600 | NP_001277636.1 |
| NM_011866.2 | 13 | 1554 | NP_035996.2 | |
| AF110507.1 | 13 | 1425 | AAD31544.1 | |
| AK039249.1 | 14 | 1600 | BAC30292.1 | |
| AK147690.1 | 12 | 1471 | ||
| AK162221.1 | 13 | 1209 | ||
| AK162804.1 | 11 | 1095 | BAE37063.1 | |
| AK166310.1 | 5 | 566 | BAE38696.1 | |
| AY360383.1 | 13 | 1554 | AAR12579.1 | |
| BC113201.1 | 13 | 1420 | AAI13202.1 |
More Information
Set Membership: |
|

Genomic Map