Assay Details
Target Gene Details
Entrez Gene ID: | 18754 |
Gene Name: | protein kinase C, epsilon |
Gene Aliases: |
5830406C15Rik, PKC[e], PKCepsilon, Pkce, R75156 |
Location: |
Chr.17:86166971-86657919 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkce | NM_011104.3 | 7 | 1694 | NP_035234.1 |
| AF028009.1 | 7 | 895 | AAB84189.1 | |
| AF325507.1 | 7 | 886 | AAG53692.1 | |
| AK082438.1 | 7 | 1695 | ||
| AK085351.1 | 8 | 1106 |
More Information
Set Membership: |
|

Genomic Map