Assay Details
Target Gene Details
Entrez Gene ID: | 98053 |
Gene Name: | general transcription factor IIF, polypeptide 1 |
Gene Aliases: |
2810405L04Rik, C76800, RAP74 |
Location: |
Chr.17:57003401-57011393 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtf2f1 | NM_133801.2 | 5 | 624 | NP_598562.1 |
| AK013002.1 | 5 | 638 | BAB28588.1 | |
| AK078048.1 | 5 | 637 | BAC37108.1 | |
| AK168236.1 | 5 | 637 | BAE40186.1 | |
| AK168243.1 | 5 | 637 | BAE40193.1 | |
| BC023081.1 | 5 | 624 | AAH23081.1 | |
| BC031123.1 | 5 | 754 | AAH31123.1 |
More Information
Set Membership: |
|

Genomic Map