Assay Details
Target Gene Details
Entrez Gene ID: | 56771 |
Gene Name: | mediator complex subunit 20 |
Gene Aliases: |
1110011O05Rik, 2410115I17Rik, AU018348, Trfp |
Location: |
Chr.17:47611596-47624418 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med20 | NM_020048.3 | NP_064432.1 | ||
| AJ245617.1 | CAB53514.1 | |||
| AK041624.1 | 1 | 60 | ||
| AK043396.1 | BAC31536.1 | |||
| AK146345.1 | BAE27098.1 | |||
| BC060122.1 | AAH60122.1 |
More Information
Set Membership: |
|

Genomic Map