Assay Details
Target Gene Details
Entrez Gene ID: | 225010 |
Gene Name: | lysocardiolipin acyltransferase 1 |
Gene Aliases: |
AI181996, Agpat8, Alcat1, Gm91, Lycat |
Location: |
Chr.17:73107985-73243368 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lclat1 | NM_001081071.2 | 4 | 626 | NP_001074540.1 |
| NM_001177967.1 | 4 | 605 | NP_001171438.1 | |
| NM_001177968.1 | 4 | 594 | NP_001171439.1 | |
| AK144580.1 | 4 | 569 | BAE25946.1 | |
| BC147499.1 | 4 | 544 | ||
| BC150884.1 | 4 | 533 | ||
| BC158076.1 | 4 | 565 | ||
| BC158079.1 | 4 | 565 |
More Information
Set Membership: |
|

Genomic Map