Assay Details
Target Gene Details
Entrez Gene ID: | 76890 |
Gene Name: | mediator of cell motility 1 |
Gene Aliases: |
0610016J10Rik, D930048L02Rik |
Location: |
Chr.17:74198398-74295544 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Memo1 | NM_133771.2 | NP_598532.1 | ||
| AF363447.1 | AAL34463.1 | |||
| AK159560.1 | BAE35184.1 | |||
| BC013819.1 | AAH13819.1 | |||
| BF466279.1 |

Genomic Map