Assay Details
Target Gene Details
Entrez Gene ID: | 69191 |
Gene Name: | protein disulfide isomerase associated 2 |
Gene Aliases: |
1810041F13Rik, AI661267, Pdip, Pdipl |
Location: |
Chr.17:26195999-26199132 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdia2 | NM_001081070.1 | NP_001074539.1 | ||
| AK007743.1 | ||||
| BC116671.1 |
More Information
Set Membership: |
|

Genomic Map