Assay Details
Target Gene Details
Entrez Gene ID: | 16867 |
Gene Name: | luteinizing hormone/choriogonadotropin receptor |
Gene Aliases: |
Gpcr19-rs1, LH-R, Lhr |
Location: |
Chr.17:88716488-88792059 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lhcgr | NM_013582.2 | NP_038610.1 | ||
| AK030325.1 | ||||
| AK054331.1 | ||||
| AK138461.1 | ||||
| M81310.1 | AAA39432.1 |
More Information
Set Membership: |
|

Genomic Map