Assay Details
Target Gene Details
Entrez Gene ID: | 58230 |
Gene Name: | ring finger protein 8 |
Gene Aliases: |
3830404E21Rik, AIP37 |
Location: |
Chr.17:29614831-29641659 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf8 | NM_021419.1 | 1 | 82 | NP_067394.1 |
| AK076180.1 | 1 | 62 | BAC36236.1 | |
| AK147168.1 | 1 | 104 | BAE27732.1 | |
| BC021778.1 | 1 | 82 | AAH21778.1 |
More Information
Set Membership: |
|

Genomic Map