Assay Details
Target Gene Details
Entrez Gene ID: | 58230 |
Gene Name: | ring finger protein 8 |
Gene Aliases: |
3830404E21Rik, AIP37 |
Location: |
Chr.17:29614831-29641659 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf8 | NM_021419.1 | NP_067394.1 | ||
| AK076180.1 | BAC36236.1 | |||
| AK147168.1 | BAE27732.1 | |||
| BC021778.1 | AAH21778.1 | |||
| CT010295.1 | CAJ18503.1 |
More Information
Set Membership: |
|

Genomic Map