Assay Details
Target Gene Details
Entrez Gene ID: | 18607 |
Gene Name: | 3-phosphoinositide dependent protein kinase 1 |
Gene Aliases: |
Pdk1 |
Location: |
Chr.17:24073680-24150922 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdpk1 | NM_001080773.2 | 10 | 1146 | NP_001074242.1 |
| NM_001286662.1 | 10 | 1064 | NP_001273591.1 | |
| NM_011062.4 | 10 | 1146 | NP_035192.2 | |
| AF079535.1 | 10 | 1103 | AAC96115.1 | |
| AF086625.1 | 10 | 1112 | AAC67544.1 | |
| AF126294.1 | 10 | 1087 | AAD38505.1 | |
| AK147149.1 | 10 | 1138 | BAE27716.1 | |
| AK147835.1 | 10 | 1121 | BAE28171.1 | |
| AK149162.1 | 10 | 1080 | ||
| AK149289.1 | 10 | 1104 | BAE28793.1 | |
| AK162677.1 | 10 | 1106 | BAE37017.1 | |
| AK165564.1 | 10 | 1083 | ||
| AK188106.1 | 2 | 114 | ||
| AK193731.1 | 2 | 79 | ||
| AK205439.1 | 1 | 59 | ||
| AK219503.1 | 2 | 72 | ||
| AY062008.1 | 10 | 1042 | AAL47185.1 | |
| BC146001.1 | 10 | 1083 | ||
| BC146003.1 | 10 | 1083 |
More Information
Set Membership: |
|

Genomic Map