Assay Details
Target Gene Details
Entrez Gene ID: | 18607 |
Gene Name: | 3-phosphoinositide dependent protein kinase 1 |
Gene Aliases: |
Pdk1 |
Location: |
Chr.17:24073680-24150922 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdpk1 | NM_001080773.2 | NP_001074242.1 | ||
| NM_001286662.1 | NP_001273591.1 | |||
| NM_011062.4 | NP_035192.2 | |||
| AF079535.1 | AAC96115.1 | |||
| AF086625.1 | AAC67544.1 | |||
| AF126294.1 | AAD38505.1 | |||
| AK147149.1 | BAE27716.1 | |||
| AK147835.1 | BAE28171.1 | |||
| AK149162.1 | ||||
| AK149289.1 | BAE28793.1 | |||
| AK162677.1 | BAE37017.1 | |||
| AK165564.1 | ||||
| AK188106.1 | ||||
| AK193731.1 | ||||
| AK205439.1 | ||||
| AK215448.1 | ||||
| AK219503.1 | ||||
| AY062008.1 | AAL47185.1 | |||
| BC146001.1 | ||||
| BC146003.1 |
More Information
Set Membership: |
|

Genomic Map