Assay Details
Target Gene Details
Entrez Gene ID: | 56716 |
Gene Name: | MTOR associated protein, LST8 homolog (S. cerevisiae) |
Gene Aliases: |
0610033N12Rik, AA409454, AI505104, AI851821, Gbl |
Location: |
Chr.17:24473550-24479114 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mlst8 | NM_001252463.1 | NP_001239392.1 | ||
| NM_001252464.1 | NP_001239393.1 | |||
| NM_001252465.1 | NP_001239394.1 | |||
| NM_019988.5 | NP_064372.2 | |||
| AF237676.1 | AAF37719.1 | |||
| AK002751.1 | BAB22328.1 | |||
| AK038515.1 | BAC30024.1 | |||
| AK040100.1 | BAC30510.1 | |||
| AK048102.1 | BAC33243.1 | |||
| AK077680.1 | BAC36952.1 | |||
| AK083731.1 | BAC39006.1 | |||
| AK088951.1 | ||||
| BC015279.1 | AAH15279.1 | |||
| BM947104.1 | ||||
| CF750475.1 |

Genomic Map