Assay Details
Target Gene Details
Entrez Gene ID: | 14979 |
Gene Name: | H2-K region expressed gene 6 |
Gene Aliases: |
D17H6S112E, H-2Ke6, Hsd17b8, Ke-6, Ke6, Ring2 |
Location: |
Chr.17:34026033-34028055 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2-Ke6 | NM_013543.2 | 9 | 915 | NP_038571.2 |
| BC086927.1 | 9 | 915 | AAH86927.1 |
More Information
Set Membership: |
|

Genomic Map