Assay Details
Target Gene Details
Entrez Gene ID: | 56462 |
Gene Name: | mitochondrial carrier homolog 1 (C. elegans) |
Gene Aliases: |
2310034O17Rik, AI255158, AU018396, C77849, PSAP |
Location: |
Chr.17:29332076-29347927 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtch1 | NM_019880.3 | NP_063933.1 | ||
| AF176007.2 | AAD52645.2 | |||
| AF192558.2 | AAF12792.2 | |||
| AK009623.1 | BAC25262.1 | |||
| AK154010.1 | BAE32313.1 | |||
| AK178745.1 | ||||
| AK178762.1 | ||||
| AK183124.1 | ||||
| AK195241.1 | ||||
| AK199429.1 | ||||
| AK200271.1 | ||||
| AK201694.1 | ||||
| AK207873.1 | ||||
| AK208166.1 | ||||
| AK210039.1 | ||||
| AK214576.1 | ||||
| BC023135.1 | AAH23135.1 | |||
| BC026436.1 | AAH26436.1 | |||
| BC027274.1 | AAH27274.1 |
More Information
Set Membership: |
|

Genomic Map