Assay Details
Target Gene Details
Entrez Gene ID: | 18999 |
Gene Name: | POU domain, class 5, transcription factor 1 |
Gene Aliases: |
NF-A3, Oct-3, Oct-3/4, Oct-4, Oct3, Oct3/4, Oct4, Otf-3, Otf-4, Otf3, Otf3-rs7, Otf3g, Otf4 |
Location: |
Chr.17:35506032-35510777 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pou5f1 | NM_001252452.1 | NP_001239381.1 | ||
| NM_013633.3 | NP_038661.2 | |||
| AB221654.1 | ||||
| AB375269.1 | ||||
| AB375270.1 | ||||
| AB375271.1 | ||||
| AB375272.1 | ||||
| AB375273.1 | ||||
| AB375276.1 | ||||
| AB375277.1 | ||||
| AB375278.1 | ||||
| AK145321.1 | ||||
| BC068268.1 | AAH68268.1 | |||
| HM346526.1 | ||||
| M34381.1 | AAA39844.1 | |||
| X52437.1 | CAA36682.1 |

Genomic Map