Assay Details
Target Gene Details
Entrez Gene ID: | 67097 |
Gene Name: | ribosomal protein S10 |
Gene Aliases: |
2210402A09Rik |
Location: |
Chr.17:27630420-27635480 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps10 | NM_025963.3 | NP_080239.1 | ||
| AK002558.1 | ||||
| AK008797.1 | BAB25901.1 | |||
| AK011065.1 | BAB27372.1 | |||
| AK020219.1 | ||||
| AK136533.1 | BAE23033.1 | |||
| AK145441.1 | BAE26437.1 | |||
| AK150803.1 | BAE29866.1 | |||
| AK151703.1 | BAE30626.1 | |||
| AK157104.1 | BAE33964.1 | |||
| AK203128.1 | ||||
| BC003853.1 | AAH03853.1 | |||
| BC019725.1 | AAH19725.1 | |||
| BC086919.1 | AAH86919.1 | |||
| BC089323.1 | AAH89323.1 |
More Information
Set Membership: |
|

Genomic Map